Learn More
Bachem Angiotensin I (1-9) Trifluoroacetate
SDP
Supplier:  Bachem 4033395.0025
Encompass_Preferred
Angiotensin I (1-9) Trifluoroacetate, H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-OH, 34273-12-6, C56H78N16O13, Mr 1183.34, 25mg. Angiotensin I (1-9) is found in vivo and accumulates in animals treated with ACE inhibitors. It has been shown that the nonapeptide may be generated in heart and kidney by endothelium-associated ACE2. Kramkowski et al. observed that DRVYIHPFH generated from Ang I in platelets enhances thrombosis in rats.
Catalog No. 50-259-917