Learn More
Bachem Angiotensin I Trifluoroacetate
SDP
Supplier:  Bachem 4002346.0025
Encompass_Preferred
Angiotensin I Trifluoroacetate, H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-OH, 484-42-4, C62H89N17O14, Mr 1296.50, 25mg. Angiotensinogen (1-10), Ang I, Angiotensin I, human. Angiotensin I, DRVYIHPFHL, is cleaved by angiotensin-converting enzyme (ACE) yielding Ang II and the dipeptide His-Leu. Ang I is also converted to Ang II by chymase, especially in the human heart. Hence, this mast cell protease is also involved in blood pressure regulation. The chymase pathway gains importance in inflammatory conditions.
Catalog No. 50-194-6746