Learn More
Medchemexpress LLC HY-P1515A 5mg Medchemexpress, Angiotensin II (3-8), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1515A5MG
Encompass_Preferred
Medchemexpress, HY-P1515A 5mg Angiotensin II (3-8), human (TFA) CAS: Angiotensin II (3-8), human (TFA) is a less effective agonist at the angiotensin AT 1 receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4217