Learn More
Medchemexpress LLC Angiotensin II (3-8), human (TFA) | 99.1% | 888.93 | 10 MG

Supplier: Medchemexpress LLC HYP1515A10MG
Angiotensin II (3-8), human TFA is a less effective agonist at the angiotensin AT1 receptor. This product is provided as a white to off-white solid.
- Molecular weight: 888.93
- Purity: 99.1%
- Appearance: Solid
- Soluble in H₂O at 50 mg/mL (needs ultrasonic)
- Store powder at -80°C for 2 years or -20°C for 1 year
- Store in solvent at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.