Learn More
CPC Scientific H-Val-Tyr-Ile-His-Pro-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ANGT006A
Encompass
Hexapeptide that binds specifically with a new angiotensin binding site to distinguish and separate from angiotensin II receptors. It is also one of the major metabolites of angiotensin II.SEQUENCE: H-Val-Tyr-Ile-His-Pro-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: VYIHPFMOLECULAR FORMULA: C40H54N8O8MOLECULAR WEIGHT: 774.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [23025-68-5]SYNONYMS: RESEARCH AREA: CardiovascularREFERENCES:K. Hermann et al., J. Neurochem., 52, 863 (1989)G.N. Swanson et al., Regul. Peptides, 40, 409 (1992)SKU(s): ANGT-006A
Catalog No. 50-210-8767