Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Angled Mounting Block, 30 °
SDP
Supplier:  Electron Microscopy Sciences 5015322
Encompass
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 30°
Use: Models 150, 151, 155
Catalog No. 50-980-982