Learn More
Bellco Glass, Inc. Angled Sample Pipe Assembly ,15L For 10mm Port
SDP
Supplier:  Bellco Glass, Inc. 790915108
Encompass_Preferred
Size: 15lMaterial: 316l sst with electropolish finish - ra 32 or better - inside & outTube dimensions: 1/4" od x 5/32" id x 17 5/8" l x 2" bendInsertion length: 15"lSupply with o-ring & sst nut. MADE IN THE USA
Catalog No. 50-194-6490