Learn More
World Precision Instrument Angled Scissors, 11.5 cm, Angled, Serrated, Sharp/Sharp
SDP
Supplier:  World Precision Instrument 503225
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-252-1286