Learn More
Medchemexpress LLC HY-N7674 5mg Medchemexpress, Angoline CAS:21080-31-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN76745MG
Encompass_Preferred
Medchemexpress, HY-N7674 5mg Angoline CAS:21080-31-9 Angoline is a potent and selective IL6/STAT3 signaling pathway inhibitor with an IC50 of 11.56 μM. Angoline inhibits STAT3 phosphorylation and its target gene expression, and inhibits cancer cell proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3005