Learn More
Medchemexpress LLC Angiopoietin-2 | >95.0% | 50UG

Supplier: Medchemexpress LLC HYP70039650UG
Recombinant canine angiopoietin-2 protein expressed in HEK293 cells with a C-terminal 6xHis tag. The glycosylated protein is supplied lyophilized from a 20 mM MOPS, 150 mM NaCl, pH 7.4, 8% trehalose formulation and is intended for research use in studies of angiogenesis and endothelial signaling.
- Recombinant full-length mature protein (Y19-F495).
- Expressed in HEK293 cells to retain native glycosylation.
- Includes C-terminal 6xHis tag for purification and detection.
- Purity greater than 95% by reducing SDS-PAGE.
- Endotoxin level <1 EU/μg by LAL method.
- Lyophilized formulation with trehalose for stability.
- Store at -20°C; aliquot and store at -80°C for extended storage.
- Suitable for angiogenesis and endothelial cell signaling research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.