Learn More
Genscript Corporation ANGPTL3/Angiopoietin-like 3Bi
Supplier:  Genscript Corporation Z04398100
Encompass
ANGPTL3 is a secreted glycoprotein that is structurally related to the angiopoietins Mature human ANGPTL3 contains an N-terminal coiled coil domain and a C terminal fibrinogen-like domain ANGPTL3 is expressed in the liver from early in development through adulthood Acts in part as a hepatokine that is involved in regulation of lipid and glucose metabolism Proposed to play a role in the trafficking of energy substrates to either storage or oxidative tissues in response to food intake
Catalog No. 50-000-58856