Learn More
Selleck Chemical LLC Angstrom6-5mg
SDP
Supplier:  Selleck Chemical LLC P11615MG
Encompass_Preferred
Angstrom6 (A6 Peptide) is a capped, eight l-amino acid peptide (Ac-KPSSPPEE-NH2) derived from the biologically active connecting peptide domain of the serine protease, human urokinase plasminogen activator (uPA), which can binds to CD44 resulting in the inhibition of migration, invasion, and metastasis of tumor cells, and the modulation of CD44-mediated cell signaling.
Catalog No. 50-238-9886