Learn More
Medchemexpress LLC Anguizole | 442666-98-0 | 99.5% | C17H11ClF2N4O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY1332110MG
Encompass_Preferred
Anguizole is a small-molecule inhibitor of hepatitis C virus (HCV) replication that alters NS4B subcellular distribution. Supplied as a light yellow solid for laboratory research, it offers high purity and documented storage stability for biochemical and cell-based studies.
  • Inhibits HCV replication and alters NS4B localization.
  • High purity (99.5%) suitable for biochemical and cell-based assays.
  • Molecular formula C17H11ClF2N4O2S; molecular weight 408.81.
  • Available in multiple pack sizes for flexible experiment planning.
  • Stable when stored as powder at -20°C; solvent stability documented.
Catalog No. 50-002-61330