Learn More
Medchemexpress LLC Anhydrotetracycline hydrochloride | 13803-65-1 | MFCD00151453 | >98% | 462.88 | C22H23ClN2O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11866025MG
Encompass_Preferred
Anhydrotetracycline hydrochloride is a tetracycline biosynthetic precursor supplied as an analytical standard for research and analytical applications. It acts as a potent competitive inhibitor of tetracycline destructase enzymes and is commonly used as an effector in tetracycline-controlled gene expression systems and in HPLC/GC/MS method development.
  • Analytical standard grade suitable for qualitative and quantitative analysis.
  • Potent competitive inhibitor of tetracycline destructase enzymes.
  • Functions as an effector for tetracycline-controlled gene expression systems.
  • Molecular weight 462.88 and formula C22H23ClN2O7.
  • Available in multiple package sizes, including 25 mg.
Catalog No. 50-003-08189