Learn More
Medchemexpress LLC Anhydrotetracycline hydrochloride | 13803-65-1 | MFCD00151453 | 99.7% | 462.88 | C22H23ClN2O7 | 50 MG

Supplier: Medchemexpress LLC HY11866050MG
Anhydrotetracycline hydrochloride is the hydrochloride salt of anhydrotetracycline, a tetracycline-family compound supplied as a research reagent for biochemical and molecular biology studies. It acts as an effector in tetracycline-dependent gene expression systems and as an inhibitor in tetracycline-destructase enzyme studies. The material is provided as a yellow to orange crystalline solid suitable for regulator-binding assays and enzymology.
- Purity approximately 99.7%.
- Molecular weight 462.88 g/mol.
- Chemical formula C22H23ClN2O7.
- Physical form solid, yellow to orange.
- Available in small research sizes (5-500 mg) and solution forms.
- Typical applications regulator-binding assays and tetracycline-related enzymology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.