Learn More
Medchemexpress LLC ANI-7 100mg | 931417-26-4 | 100 MG

Supplier: Medchemexpress LLC HY117102100MG
ANI-7 ((Z)-2-(3,4-dichlorophenyl)-3-(1H-pyrrol-2-yl)acrylonitrile) is a small-molecule activator of the aryl hydrocarbon receptor (AhR) supplied as a solid for research use. It is intended for use in cell-based and biochemical assays to study AhR signaling, CYP1 enzyme induction, and related cancer biology mechanisms.
- Activator of aryl hydrocarbon receptor for research applications.
- High purity (~99.01%) suitable for biochemical and cellular assays.
- CAS number 931417-26-4 for unambiguous identification.
- Solid, light yellow to yellow appearance; soluble in DMSO for assay preparation.
- Molecular weight 263.12 g/mol, useful for dosing and calculations.
- Available in milligram-scale quantities for screening and preclinical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.