Learn More
Medchemexpress LLC Recombinant human bone morphogenetic protein 15 (BMP-15), His-tagged | ≥98.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP700024AF50UG
Encompass_Preferred
Recombinant human bone morphogenetic protein 15 (BMP-15) is a His-tagged growth factor produced in Escherichia coli and supplied lyophilized for research use in cell culture and biochemical assays. The product is provided with handling guidance for reconstitution and storage to preserve activity and integrity.
  • His-tagged recombinant human BMP-15 expressed in E. coli.
  • Lyophilized formulation for convenient storage and transport.
  • High purity as determined by reducing SDS-PAGE (≥98.0%).
  • Approximate molecular weight 13-18 kDa based on SDS-PAGE.
  • Recommended reconstitution at 100-200 μg/mL in ddH2O.
  • Stable long-term when aliquoted and stored at -20°C or -80°C with carrier protein.
Catalog No. 50-001-77445