Learn More
Medchemexpress LLC Animal-free BMP-3 protein, human (His) | 95.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP700026AF10UG
Encompass_Preferred
Recombinant human bone morphogenetic protein 3 (BMP-3) with a C-terminal His tag, expressed in E. coli and supplied as an animal-free recombinant protein for research and cell culture use. The preparation corresponds to the mature BMP-3 region (residues Q363-R472) and is provided in small laboratory quantities suitable for in vitro assays.
  • Animal-free recombinant human BMP-3 with C-terminal His tag.
  • Expressed in E. coli for consistent, non-glycosylated production.
  • Corresponds to the mature BMP-3 sequence (residues Q363-R472).
  • Provided in small laboratory quantity suitable for cell culture and in vitro assays.
  • High purity suitable for biological assays (>95% reported by distributor).
Catalog No. 50-001-78306