Learn More
Medchemexpress LLC Epidermal growth factor, human recombinant (C-terminal 6xHis tag) | >95.0% | 2UG
SDP
Supplier:  Medchemexpress LLC HYP700051AF2UG
Encompass_Preferred
Recombinant human epidermal growth factor (EGF) produced without animal-derived materials. The protein is expressed in Escherichia coli and supplied as a lyophilized powder with a C-terminal 6xHis tag. It is characterized by high purity and potent biological activity suitable for cell-based assays and research applications, with recommended reconstitution and storage conditions provided by the manufacturer.
  • Produced without animal-derived materials.
  • Expressed in Escherichia coli with a C-terminal 6xHis tag.
  • Supplied as lyophilized powder for long-term stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 9-11 kDa.
  • High biological activity; ED50 ~0.736 ng/mL in cell proliferation assays.
  • Recommended reconstitution 100-200 μg/mL; store at -20°C or colder.
Catalog No. 50-001-78342