Learn More
Medchemexpress LLC Animal-Free FGF-21 10ug
SDP
Supplier:  Medchemexpress LLC HYP70473AF10UG
Encompass_Preferred
Recombinant animal-free human Fibroblast Growth Factor 21 (FGF-21) with a C-terminal His tag, expressed in E. coli and supplied as a lyophilized powder for research use. It supports studies of metabolic regulation, insulin sensitivity, and FGFR/KLB signaling, and is provided in a small vial suitable for biochemical assays and cell-based experiments.
  • Animal-free recombinant human protein.
  • C-terminal His tag for purification and detection.
  • Expressed in E. coli for high-yield production.
  • Lyophilized formulation for stability and long-term storage.
  • Suitable for biochemical assays and cell-based studies.
  • Available in a 10 μg vial for low-volume experiments.
Catalog No. 50-001-85494