Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free GDNF protein, mouse (His) | ≥98.0% | 2UG
SDP

Supplier:  Medchemexpress LLC HYP700179AF2UG

Encompass_Preferred

Recombinant, animal-free mouse glial cell line-derived neurotrophic factor (GDNF) produced in E. coli with a C-terminal His tag. Supplied for cell culture research, this neurotrophic protein promotes survival of dopaminergic neurons and is provided as a lyophilized preparation with recommended storage at -20 °C.

  • Promotes survival of dopaminergic neurons.
  • Recombinant, animal-free protein suitable for research.
  • Produced in E. coli expression system.
  • Consists of 134 amino acids (S78-I211).
  • C-terminal His tag for detection and purification.
  • Purity ≥98.0% (as reported for specific batches).
  • Store at -20 °C; shelf life reported up to 2 years.
  • Available in small research sizes suitable for cell culture.

Catalog No. 50-001-78206


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.