Learn More
Medchemexpress LLC Interleukin-10 (mouse), recombinant (His-tag) | 130068-27-8 | 95.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP700188AF50UG
Encompass_Preferred
Recombinant mouse interleukin-10 (IL-10) is supplied as an N-terminal His-tagged protein expressed in E. coli using animal-free production methods for research applications. The preparation has an approximate molecular weight of 19.69 kDa and is provided at high purity (~95%) in small research-scale quantities; it is shipped on cold packs to maintain stability.
Catalog No. 50-001-77593