Learn More
Medchemexpress LLC Recombinant human interleukin-11 (His-tag) | 145941-26-0 | >95.0% | 2UG
SDP
Supplier:  Medchemexpress LLC HYP700098AF2UG
Encompass_Preferred
Recombinant human interleukin-11 (His-tag) produced without animal-derived components and supplied as a carrier-free preparation for in vitro research. IL-11 is a cytokine that supports hematopoietic and megakaryocyte proliferation; this recombinant form is intended for use in biochemical and cell-based assays, and is characterized by high purity and compatibility with common detection methods.
  • Animal-free production suitable for serum-free workflows.
  • His-tag enables affinity purification and straightforward detection.
  • Carrier-free formulation minimizes background in downstream assays.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Suitable for western blot, ELISA, and intracellular flow cytometry.
Catalog No. 50-001-77566