Learn More
Medchemexpress LLC Recombinant mouse interleukin-15 (His-tag) | >95.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP700193AF50UG
Encompass_Preferred
Recombinant mouse interleukin-15 (IL-15) is an animal-free, His-tagged cytokine produced in Escherichia coli and supplied for cell culture research applications. The protein corresponds to the mature mouse IL-15 sequence (UniProt P48346, residues N49-S162) and is provided as a small-scale vial for in-lab assays.
  • Animal-free recombinant IL-15 with N-terminal His tag.
  • Produced in Escherichia coli for consistent lot-to-lot supply.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 14.06 kDa.
  • Supplied in a 50 μg vial suitable for cell culture use.
  • Sequence corresponds to UniProt P48346 residues N49-S162.
Catalog No. 50-001-77769