Learn More
Medchemexpress LLC Animal-Free IL-16 M 10ug
SDP
Supplier:  Medchemexpress LLC HYP72590AF10UG
Encompass_Preferred
IL-16 is a modulator in inflammatory processes and tumorigenesis IL16 can bind to the CD4 molecule and activate monocytes and stimulates the secretion of inflammatory cytokines Besides IL16 can induce expression of IL2R and and synergize with IL2 to augment CD4 T cell activation and proliferation IL16 can induce migration in CD4 lymphocytes monocytes and eosinophils as a chemoattractant factor[1][2][3] Animal-Free IL-16 Protein Mouse (His) is the recombinant mouse-derived animal-FreeIL-16 protein expressed by E coli with C-His labeled tag This product is for cell culture use only
Catalog No. 50-001-82536