Learn More
Medchemexpress LLC Interleukin-17F | >95.0% | 50UG

Supplier: Medchemexpress LLC HYP700195AF50UG
Recombinant animal-free interleukin-17F (IL-17F) protein, mouse sequence with a C-terminal His tag, produced in E. coli for research use. Intended for in vitro studies of cytokine signaling, inflammation, and host defense. Purity is greater than 95% by reducing SDS-PAGE; predicted molecular weight is 15.8 kDa (observed ~17-25 kDa by SDS-PAGE).
- Recombinant mouse IL-17F with C-terminal His tag.
- Produced in E. coli using animal-free production methods.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Predicted molecular weight 15.8 kDa; observed ~17-25 kDa by SDS-PAGE.
- Provided in research-scale quantity suitable for in vitro assays.
- Applications include studies of cytokine signaling, inflammation, and antimicrobial responses.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.