Learn More
Medchemexpress LLC Animal-free IL-19 protein, human (His) | 98.0% | 2UG

Supplier: Medchemexpress LLC HYP700109AF2UG
Recombinant human interleukin-19 with a C-terminal His tag, expressed in Escherichia coli and supplied as a lyophilized powder for research use in cell culture. Characterized for purity, activity, and low endotoxin for reliable bioassays.
- Species: human.
- Expression system: Escherichia coli.
- Tag: C-terminal 6xHis.
- Purity: 98.0% by reducing SDS-PAGE.
- Molecular weight: ~18 kDa.
- Recommended reconstitution: 100-200 μg/mL in ddH2O.
- Endotoxin: <0.1 EU per μg (LAL method).
- Bioactivity: ED50 <1.2 ng/mL in BaF3 proliferation assay.
- Storage: store at -20°C; stable at 4°C for 1 week after reconstitution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.