Learn More
Medchemexpress LLC Mouse interleukin-19 (IL-19), C-terminal His tag | >95.0% | 2UG

Supplier: Medchemexpress LLC HYP700196AF2UG
Recombinant mouse interleukin-19 (IL-19) is supplied as a lyophilized, C-terminally His-tagged protein produced in an E. coli expression system and intended for research use. It is provided with high purity and recommended reconstitution and storage conditions to support consistent experimental results.
- Recombinant mouse IL-19, C-terminally His-tagged.
- Expressed in E. coli for consistent production.
- Purity greater than 95% by reducing SDS-PAGE.
- Predicted molecular mass ~18.5 kDa, sequence L25-A176.
- Lyophilized formulation from PBS, pH 7.4, containing trehalose.
- Recommended reconstitution to 100-200 μg/mL in ddH2O.
- Store at -20°C; after reconstitution stable short term at 4°C or long term at -20°C/-80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.