Learn More
Medchemexpress LLC Animal-free IL-20 protein, mouse (His) | 10UG

Supplier: Medchemexpress LLC HYP700198AF10UG
Recombinant animal-free mouse interleukin-20 (IL-20) is supplied as a C-terminal His-tagged protein expressed in Escherichia coli for in vitro cell culture research. The product is provided as a 10 μg quantity and is stored at -20°C. Concentration, purity, and formulation are not specified in available distributor listings and should be confirmed with the manufacturer before use.
- Animal-free recombinant cytokine suitable for cell culture research.
- C-terminal His tag for detection and purification.
- Expressed in Escherichia coli expression system.
- Supplied as a 10 μg quantity and stored at -20°C.
- Molecular weight approximately 18.52 kDa.
- Concentration and purity not specified; verify before use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.