Learn More
Medchemexpress LLC Animal-free IL-23 p19 protein, human (His) | >95.0% | 2UG
SDP
Supplier:  Medchemexpress LLC HYP700114AF2UG
Encompass_Preferred
Recombinant human IL-23 alpha (p19) is supplied as an N-terminal 6xHis-tagged protein produced in E. coli and provided lyophilized for research use. It is intended for cell-culture and biochemical assays requiring the IL-23 p19 subunit and is characterized for consistency across batches.
  • N-terminal His-tag for affinity purification and detection.
  • Expressed in E. coli to provide consistent recombinant material.
  • Lyophilized formulation stable for storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in multiple small-scale sizes suitable for assay development.
Catalog No. 50-001-77889