Learn More
Medchemexpress LLC Animal-free IL-3 protein, mouse (His) | 10UG
SDP
Supplier:  Medchemexpress LLC HYP700205AF10UG
Encompass_Preferred
Animal-Free IL-3 Protein, Mouse (His) is a lyophilized, recombinant mouse interleukin-3 produced in an animal-free E. coli expression system and provided with a His tag. It is intended for in vitro research use (hematopoiesis, immune cell stimulation) and should be reconstituted and handled per the manufacturer's datasheet.
Catalog No. 50-001-77360