Learn More
Medchemexpress LLC Interleukin-33, mouse recombinant (His-tag) | 10UG

Supplier: Medchemexpress LLC HYP700208AF10UG
Animal-free interleukin-33 (mouse) is a recombinant cytokine expressed in E. coli with a C-terminal His tag, supplied for research and cell-culture applications. It activates NF-κB and MAPK signaling and promotes Th2-associated responses, including activation of mast cells, basophils, eosinophils, and natural killer cells. Supplied in microgram quantities for functional and signaling studies.
- Recombinant mouse IL-33 with C-terminal His tag.
- Produced in E. coli using animal-free processes.
- Activates NF-κB and MAPK signaling pathways.
- Promotes Th2-associated immune responses.
- Suitable for cell culture and functional assays.
- Molecular weight approximately 17-25 kDa.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.