Learn More
Medchemexpress LLC Interleukin-34 (IL-34) | >95.0% | 10UG

Supplier: Medchemexpress LLC HYP700125AF10UG
Recombinant human interleukin-34 (IL-34) is a His-tagged cytokine supplied lyophilized for research use. It promotes proliferation, survival, and differentiation of monocytes and macrophages and is provided with low endotoxin and high purity for reliable in vitro assays.
- High purity (>95%) as determined by reducing SDS-PAGE.
- Low endotoxin level (<0.1 EU per μg) suitable for cell-based assays.
- C-terminal His tag for detection and purification.
- Mature sequence corresponding to residues N21-P242 for functional activity.
- Lyophilized formulation for enhanced stability and convenient storage.
- Biological activity confirmed (ED50 <30 pg/mL) in PBMC proliferation assays.
- Recommended storage at -20°C and aliquot freezing for long-term use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.