Learn More
Medchemexpress LLC Animal-free IL-4 protein, pig (His) | 98.0% | 2UG

Supplier: Medchemexpress LLC HYP700248AF2UG
Animal-Free IL-4 Protein, Pig (His) is a recombinant porcine interleukin-4 expressed in E. coli and supplied as a lyophilized, His-tagged protein for in vitro research. Certificate of analysis reports an approximate molecular weight of 15 kDa, high purity, low endotoxin, and potent activity in cell proliferation assays.
- Recombinant porcine IL-4 with C-6xHis tag.
- Expressed in E. coli and supplied lyophilized for stability.
- High purity (98.0% by reducing SDS-PAGE).
- Low endotoxin level (<0.1 EU per 1 μg) suitable for cell-based assays.
- Potent biological activity (ED50 <0.6 ng/mL in TF-1 proliferation).
- Lyophilized from PBS, pH 7.4; reconstitute to 100-200 μg/mL in ddH2O.
- Stable long-term at -20°C; aliquot and store at -20°C or -80°C after reconstitution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.