Learn More
Medchemexpress LLC Animal-Free IP-10/CX 50ug
SDP
Supplier:  Medchemexpress LLC HYP700041AF50UG
Encompass_Preferred
The IP-10/CRG-2/CXCL10 protein is a pro-inflammatory cytokine involved in a variety of biological processes including chemotaxis immune cell activation growth regulation apoptosis and vasostatic regulation During viral infection IP-10 crucially stimulates immune cell activation and migration to the site of infection Animal-Free IP-10/CXCL10 Protein Human (His) is the recombinant human-derived animal-FreeIP-10/CRG-2/CXCL10 protein expressed by E coli with N-His labeled tag This product is for cell culture use only
Catalog No. 50-001-78258