Learn More
Medchemexpress LLC Animal-Free KGF-2/FG 10ug
SDP
Supplier:  Medchemexpress LLC HYP7342AF10UG
Encompass_Preferred
KGF-2/FGF-10 proteins coordinate embryonic development regulate cell proliferation and differentiation and are indispensable in branching morphogenesis This multifunctional protein may aid in wound healing Animal-Free KGF-2/FGF-10 Protein Human (His) is the recombinant human-derived animal-FreeKGF-2/FGF-10 protein expressed by E coli with C-His C-His labeled tag This product is for cell culture use only
Catalog No. 50-001-80656