Learn More
Medchemexpress LLC Leukemia inhibitory factor (mouse) | >95.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP700220AF10UG
Encompass_Preferred
Animal-free recombinant mouse leukemia inhibitory factor (LIF) is a His-tagged cytokine produced in E. coli for cell culture and research applications. The protein corresponds to the mouse LIF sequence (UniProt P09056), shows an apparent molecular weight of approximately 17-25 kDa on SDS-PAGE, and has >95% purity by reducing SDS-PAGE.
  • Recombinant mouse LIF produced in E. coli.
  • Includes a His tag for purification and detection.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight ~17-25 kDa on SDS-PAGE.
  • Intended for cell culture and research use only.
Catalog No. 50-001-77960