Learn More
Medchemexpress LLC Macrophage colony-stimulating factor (M-CSF), recombinant human | 81627-83-0 | MFCD00166591 | >95.0% | 50UG

Supplier: Medchemexpress LLC HYP700137AF50UG
Animal-Free M-CSF Protein, Human (His) is a recombinant human macrophage colony-stimulating factor produced in E. coli and supplied lyophilized with a C-terminal 6xHis tag for research and cell-culture applications. It regulates macrophage survival, differentiation, and function, and is characterized for biological activity and high purity.
- Lyophilized C-terminal 6xHis-tagged recombinant human M-CSF.
- Supplied as a 50 μg vial for research use.
- Produced in E. coli using animal-free processes.
- Predicted molecular mass 13.3 kDa; ~13-26 kDa by reducing SDS-PAGE.
- Purity >95% as determined by reducing SDS-PAGE.
- Specific activity >2.5 x 10^8 IU/mg; ED50 <1 ng/mL (NFS-60 assay).
- Formulated in PBS with trehalose; reconstitute to 100-200 μg/mL in ddH2O.
- Store at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.