Learn More
Medchemexpress LLC Recombinant mouse macrophage colony-stimulating factor (His-tagged) | 81627-83-0 | MFCD00166591 | 2UG
SDP
Supplier:  Medchemexpress LLC HYP700222AF2UG
Encompass_Preferred
Animal-Free M-CSF Protein, Mouse (His) is a recombinant mouse macrophage colony-stimulating factor supplied as a His-tagged polypeptide expressed in E. coli. The product corresponds to residues K33-P187 (155 amino acids) and is provided for research use to support studies of macrophage survival, proliferation, and differentiation. Molecular weight is approximately 17-25 kDa by SDS-PAGE.
  • Recombinant mouse M-CSF, residues K33-P187 (155 aa).
  • His-tagged for efficient purification and detection.
  • Expressed in E. coli expression system.
  • Approximate molecular weight 17-25 kDa (SDS-PAGE).
  • Intended for research use in macrophage biology and immunology studies.
Catalog No. 50-001-78010