Learn More
Medchemexpress LLC Nodal protein, human (recombinant, C-terminal His tag) | >95.0% | 2UG
SDP
Supplier:  Medchemexpress LLC HYP700025AF2UG
Encompass_Preferred
Recombinant human Nodal protein expressed in E. coli with a C-terminal 6xHis tag, supplied as an animal-free recombinant protein for cell culture research. The protein forms disulfide-linked homodimers that participate in embryonic development and mesoderm patterning.
Catalog No. 50-001-78194