Learn More
Medchemexpress LLC Animal-free TGF beta 1/TGFB1 protein, pig (His) | ≥98.0% | 50UG

Supplier: Medchemexpress LLC HYP700251AF50UG
Animal-free TGF beta 1 (TGFB1) protein, porcine, is a recombinant His-tagged cytokine supplied as a lyophilized powder formulated in 20 mM sodium citrate, 0.2 M NaCl, pH 4.5 with trehalose. Expressed in E. coli and characterized by SDS-PAGE and LAL testing, it is intended for cell culture and biochemical assays requiring active TGF-β1 signaling.
- Recombinant porcine TGF-β1 with C-terminal His tag for detection and purification.
- Supplied as lyophilized powder formulated for stability.
- High purity, ≥98.0% by reducing SDS-PAGE.
- Low endotoxin: <0.1 EU per μg by LAL assay.
- Apparent molecular weight ~11-17 kDa (reducing SDS-PAGE).
- Store at -20°C; reconstituted material stable at 4°C for 1 week or frozen for long-term storage.
- Suitable for cell culture and biochemical assays involving TGF-β signaling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.