Learn More
Apexbio Technology LLC Aniracetam 72432-10-1 5g
SDP
Supplier:  Apexbio Technology LLC B12105000
Encompass_Preferred
Aniracetam (CAS 72432-10-1) is a nootropic compound structurally related to piracetam characterized as 1-p-anisoyl-2-pyrrolidinone It has been shown to modulate cognitive function in preclinical models possibly by influencing neurotransmitter systems implicated in memory and learning In rodent studies oral administration of aniracetam (30 50 mg/kg) prevented memory acquisition deficits induced by CO exposure and reversed cognitive impairments caused by scopolamine or electroconvulsive shock Aniracetam also mitigated learning disruptions from protein synthesis inhibitors such as chloramphenicol and cycloheximide These findings support its utility in investigating mechanisms underlying cognitive disorders and potential therapeutic approaches for memory impairment
Catalog No. 50-000-87730