Learn More
Medchemexpress LLC Aniracetam | 72432-10-1 | 99.9% | 219.24 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY109325G
Encompass_Preferred
Aniracetam is an orally active neuroprotective agent with nootropic effects. It is used to research cerebral dysfunctional disorders and can prevent CO2-induced impairment of acquisition in hypercapnia model rats.
  • Potentiates ionotropic quisqualate (iQA) responses
  • Potentiates excitatory postsynaptic potentials (EPSPs)
  • Counteracts glutamate-, kainate-, or α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid-induced cell death
  • Facilitates neuroprotective response by quisqualate and trans-1-aminocyclopentane-1, 3-dicarboxylate
  • Potentiates mGluR-coupled stimulation of phospholipase C
Catalog No. 50-003-45319