Learn More
Medchemexpress LLC Anisodamine hydrobromide (6-Hydroxyhyoscyamine hydrobromide) | 55449-49-5 | 386.28 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0584A5MG
Encompass_Preferred
Anisodamine hydrobromide, also known as 6-Hydroxyhyoscyamine hydrobromide, is a belladonna alkaloid. It functions as a non-subtype-selective antagonist for both muscarinic and nicotinic cholinoceptors, and exhibits antioxidant and anti-inflammatory properties.
  • Muscarinic acetylcholine receptor antagonist
  • Nicotinic cholinoceptor antagonist
  • Antioxidant properties
  • Anti-inflammatory properties
  • Available for research use only
Catalog No. 50-003-88754