Learn More
Medchemexpress LLC Anisole-piperazine-methanone-benzothiazole-p-methylpiperidine | 906258-69-3 | 99.14% | C25H30N4O2S | 25 MG

Supplier: Medchemexpress LLC 25MGHY145766
Anisole-piperazine-methanone-benzothiazole-p-methylpiperidine is an active compound intended for research use only. It appears as a white to off-white solid and has a molecular weight of 450.60. The compound can be dissolved in DMSO for in vitro experiments and in specific formulations for in vivo studies.
- Active compound
- For research use only
- White to off-white solid appearance
- Molecular weight of 450.60
- Soluble in DMSO for in vitro use
- Dissolution protocols for in vivo experiments
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.