Learn More
Selleck Chemical LLC Anisomycin 10mg 22862-76-6 Flagecidin
SDP
Supplier:  Selleck Chemical LLC S740910MG
Encompass_Preferred
Anisomycin (Flagecidin) is a bacterial antibiotic isolated from Streptomyces griseolus, which inhibits protein synthesis, and also act as a JNK activator. Anisomycin upregulates autophagy and increases apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4742