Learn More
Alkali Scientific Anisomycin from streptomyces griseolus [2 [(4 Methoxyphenyl)methyl] 3, 4 pyrrolidinediol 3 acetate], 10 Mg
SDP
Supplier:  Alkali Scientific ANI001CP
Encompass
Anisomycin is a protein synthesis inhibitor derived from Streptomyces species and widely used in molecular and cellular biology research. It functions by binding to the 60S ribosomal subunit, inhibiting peptide bond formation and blocking translation. This activity also induces cellular stress responses and activates signaling pathways such as MAP kinase cascades. Researchers use anisomycin to study gene expression, apoptosis, and stress related cellular mechanisms. It is commonly applied in neuroscience, cancer research, and signal transduction studies. Supplied in a 10 mg quantity, it is suitable for precise laboratory experiments requiring controlled inhibition of protein synthesis.
Catalog No. 50-006-61067