Learn More
American Research Products Inc Anti-Adeno-associated virus (AAV2), intact particles, clone A20, 50ug
Supplier:  American Research Products Inc 61055
Encompass
For characterization of different stages of infection and very useful for the analysis of the AAV assembly process. The antibody cannot be used for immunoblotting. Specifically detects intact virus particles, both empty and full capsids. With AAV-2 and AAV-3 A20 recognizes a conformational epitope of assembled capsids, not present in denatured and native unassembled capsid proteins.
Catalog No. NC1680057