Learn More
Aviva Systems Biology Corp ANKRD2 Antibody - N-terminal region (ARP42559_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42559_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANKRD2. Alternate names: ARPP, MGC104314ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58266