Learn More
Aviva Systems Biology Corp ANKS3 Antibody - N-terminal region (ARP52561_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52561_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANKS3. Alternate names: FLJ32345, FLJ32767, KIAA1977ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53697